KEY092592869 Key Organics / BIONET

1-(3-aminophenyl)-2-(3,5-dimethyl-1H-pyrazol-1-yl)ethan-1-one

  • MDL number MFCD03617767
  • Cas Number : 956206-44-3
  • Molecular Weight : 229.283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140 - 143
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93995

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9T-0803_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart