KEY128003613 Key Organics / BIONET

2-(3-chloropropyl)-3-(3,4-dimethoxyphenyl)-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid

  • MDL number MFCD09152692
  • Cas Number : 1011412-52-4
  • Molecular Weight : 403.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6686

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-0266_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart