KEY128003616 Key Organics / BIONET

4-[4-(trifluoromethyl)pyridin-2-yl]benzaldehyde

  • MDL number MFCD09607891
  • Cas Number : 952182-74-0
  • Molecular Weight : 251.208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66 - 68
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7384

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11Z-0715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart