KEY128003618 Key Organics / BIONET

4-(2-hydroxyethyl)-3-methyl-5-oxo-2,5-dihydro-1H-pyrazole-1-carbothioamide

  • MDL number MFCD09027087
  • Cas Number : 850497-29-9
  • Molecular Weight : 201.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167 - 168
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63108

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12Z-0221_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart