KEY128003628 Key Organics / BIONET

tert-butyl N-[(dimethylcarbamoyl)methyl]carbamate

  • MDL number MFCD09152695
  • Cas Number : 72287-76-4
  • Molecular Weight : 202.254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66 - 68
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88249

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7Y-0621_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart