KEY128003632 Key Organics / BIONET

benzyl N-[(benzenesulfonyl)(2H-1,3-benzodioxol-5-yl)methyl]carbamate

  • MDL number MFCD09817425
  • Cas Number : 952182-82-0
  • Molecular Weight : 425.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162 - 164
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94424

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0019_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart