KEY128003634 Key Organics / BIONET

1-benzyl 2-tert-butyl 1,2-diazinane-1,2-dicarboxylate

  • MDL number MFCD09152697
  • Cas Number : 154972-38-0
  • Molecular Weight : 320.389

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94429

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0219_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart