KEY128003638 Key Organics / BIONET

2-[4-(trifluoromethoxy)phenoxy]acetonitrile

  • MDL number MFCD09152763
  • Cas Number : 952182-87-5
  • Molecular Weight : 217.147

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119 @ 12 mBar
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94434

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0604_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart