KEY128003642 Key Organics / BIONET

4-(naphthalen-2-yl)-6H,7H-[1,2]oxazolo[3,4-d]pyridazin-7-one

  • MDL number MFCD09152703
  • Cas Number : 952182-91-1
  • Molecular Weight : 263.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260 - 262
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94439

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0708_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart