KEY128003643 Key Organics / BIONET

4-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)benzaldehyde

  • MDL number MFCD09152704
  • Cas Number : 952182-92-2
  • Molecular Weight : 254.285

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94 - 96
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94440

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0709_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart