KEY128003645 Key Organics / BIONET

4-cyclopropyl-6H,7H-[1,2]oxazolo[3,4-d]pyridazin-7-one

  • MDL number MFCD09152706
  • Cas Number : 952182-94-4
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 210 - 212
Iupac Name 4-cyclopropylisoxazolo[3,4-d]pyridazin-7(6H)-one
Inchi Code 1S/C8H7N3O2/c12-8-7-5(3-13-11-7)6(9-10-8)4-1-2-4/h3,9H,1-2H2,(H,10,12)
Inchi Key OFPMWKCRBSMJDH-UHFFFAOYSA-N

Description

Melting Point 210 - 212
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94443

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AB-0712_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart