KEY128003646 Key Organics / BIONET

ethyl 4-cyclopropanecarbonyl-1,2-oxazole-3-carboxylate

  • MDL number MFCD09152707
  • Cas Number : 952182-95-5
  • Molecular Weight : 209.201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57 - 59
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94444

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0713_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart