KEY128003647 Key Organics / BIONET

(2E)-1-(1,3-benzoxazol-7-yl)-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD09152708
  • Cas Number : 952182-96-6
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103 - 105
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94445

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0716_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart