KEY128003648 Key Organics / BIONET

1-(2-ethyl-1,3-benzoxazol-7-yl)ethan-1-one

  • MDL number MFCD09152709
  • Cas Number : 952182-97-7
  • Molecular Weight : 189.214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42 - 44
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94446

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0717_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart