KEY128003649 Key Organics / BIONET

1-(2-methyl-1,3-benzoxazol-7-yl)ethan-1-one

  • MDL number MFCD09152710
  • Cas Number : 952182-98-8
  • Molecular Weight : 175.187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 -100
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94447

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AB-0718_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart