KEY128003650 Key Organics / BIONET

5-nitro-2-(prop-2-yn-1-ylsulfanyl)pyridine

  • MDL number MFCD09152764
  • Cas Number : 77867-13-1
  • Molecular Weight : 194.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102 - 104
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94449

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0720_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart