KEY128003652 Key Organics / BIONET

2-chloro-N-(2-hydroxyethyl)-4-(piperidin-1-yl)benzamide

  • MDL number MFCD09152713
  • Cas Number : 952183-00-5
  • Molecular Weight : 282.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 100
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94454

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0726_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart