KEY128003656 Key Organics / BIONET

(E)-N'-(1H-indazol-6-yl)-N,N-dimethylethanimidamide

  • MDL number MFCD09152717
  • Cas Number : 952183-02-7
  • Molecular Weight : 202.261

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232 - 233
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94462

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0805_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart