KEY128003672 Key Organics / BIONET

ethyl 5-chloropyridine-2-carboxylate

  • MDL number MFCD07772018
  • Cas Number : 128072-93-5
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 47 - 50
Iupac Name ethyl 5-chloro-2-pyridinecarboxylate
Inchi Code 1S/C8H8ClNO2/c1-2-12-8(11)7-4-3-6(9)5-10-7/h3-5H,2H2,1H3
Inchi Key BEBBWTPLBRZIOF-UHFFFAOYSA-N

Description

Melting Point 47 - 50
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94932

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BB-0606_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart