KEY128003693 Key Organics / BIONET

2-(5-cyano-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)acetic acid

  • MDL number MFCD09607899
  • Cas Number : 56673-29-1
  • Molecular Weight : 195.134

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236 - 239
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94967

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BB-0812_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart