KEY128003717 Key Organics / BIONET

N-(2-hydroxyethyl)-4-(1H-imidazol-1-yl)benzamide

  • MDL number MFCD09607920
  • Cas Number : 952183-34-5
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 175 - 177
Iupac Name N-(2-hydroxyethyl)-4-(1H-imidazol-1-yl)benzamide
Inchi Code 1S/C12H13N3O2/c16-8-6-14-12(17)10-1-3-11(4-2-10)15-7-5-13-9-15/h1-5,7,9,16H,6,8H2,(H,14,17)
Inchi Key IVCZEZZYMPELQR-UHFFFAOYSA-N

Description

Melting Point 175 - 177
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95582

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CB-0743_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart