KEY128003718 Key Organics / BIONET

6-(dimethylamino)pyridine-3-carbaldehyde

  • MDL number MFCD08272100
  • Cas Number : 149805-92-5
  • Molecular Weight : 150.181

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57 - 59
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95584

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CB-0745_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart