KEY128003721 Key Organics / BIONET

6-(methylamino)pyridine-3-carbaldehyde

  • MDL number MFCD09607923
  • Cas Number : 72087-21-9
  • Molecular Weight : 136.154

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124 - 126
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95588

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CB-0750_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart