KEY128003742 Key Organics / BIONET

3-methyl-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09607943
  • Cas Number : 116834-96-9
  • Molecular Weight : 133.154

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158 - 160
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96035

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DB-0603_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart