KEY128003743 Key Organics / BIONET

ethyl 4-cyano-3-fluorobenzoate

  • MDL number MFCD09607944
  • Cas Number : 952183-53-8
  • Molecular Weight : 193.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-cyano-3-fluorobenzoate
Inchi Code 1S/C10H8FNO2/c1-2-14-10(13)7-3-4-8(6-12)9(11)5-7/h3-5H,2H2,1H3
Inchi Key OEYSQQVQGGLOQP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96036

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/DB-0604_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart