KEY128003745 Key Organics / BIONET

methyl 3-(ethylamino)-4-nitrobenzoate

  • MDL number MFCD09607946
  • Cas Number : 952183-54-9
  • Molecular Weight : 224.216

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92 - 94
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Orange Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96041

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/DB-0700_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart