KEY128003781 Key Organics / BIONET

2-amino-6,8-dibromoquinoline-3-carboxamide

  • MDL number MFCD08689708
  • Cas Number : 937601-68-8
  • Molecular Weight : 344.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 283 - 286
Iupac Name 2-amino-6,8-dibromo-3-quinolinecarboxamide
Inchi Code 1S/C10H7Br2N3O/c11-5-1-4-2-6(10(14)16)9(13)15-8(4)7(12)3-5/h1-3H,(H2,13,15)(H2,14,16)
Inchi Key MMDYMXKTESCOCM-UHFFFAOYSA-N

Description

Melting Point 283 - 286
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96720

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/FA-0716_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart