KEY128003797 Key Organics / BIONET

methyl 7-bromo-1-benzothiophene-2-carboxylate

  • MDL number MFCD09027095
  • Cas Number : 550998-53-3
  • Molecular Weight : 271.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99 - 102
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96807

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FA-0933_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart