KEY128003813 Key Organics / BIONET

methyl 4-[4-(hydroxymethyl)piperidin-1-yl]benzoate

  • MDL number MFCD08689747
  • Cas Number : 392690-98-1
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111 - 113
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97192

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GA-0719_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart