KEY128003834 Key Organics / BIONET

7-bromo-2,3-dihydro-1-benzofuran-5-carbaldehyde

  • MDL number MFCD08689769
  • Cas Number : 875551-14-7
  • Molecular Weight : 227.057

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109 - 111
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97235

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GA-0836_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart