KEY128003843 Key Organics / BIONET

ethyl 2-amino-4,5,6,7-tetrahydro-1,3-benzothiazole-4-carboxylate

  • MDL number MFCD08689777
  • Cas Number : 76263-11-1
  • Molecular Weight : 226.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173 - 174
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97247

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GA-0858_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart