KEY128003853 Key Organics / BIONET

2-chloro-4,5,6,7-tetrahydro-1,3-benzothiazole

  • MDL number MFCD08689787
  • Cas Number : 26846-98-0
  • Molecular Weight : 173.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 82 @ 0.1 mm Hg
Iupac Name 2-chloro-4,5,6,7-tetrahydro-1,3-benzothiazole
Inchi Code 1S/C7H8ClNS/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H2
Inchi Key LDJUCUDNAZQFIZ-UHFFFAOYSA-N

Description

Melting Point 82 @ 0.1 mm Hg
Purity 95%
Storage temp. Store under N2
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97261

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/GA-0920_SDS.pdf
hcodes H302,H312,H314,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart