KEY128003855 Key Organics / BIONET

4-bromo-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD03407342
  • Cas Number : 5194-37-6
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97264

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GA-0924_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart