KEY128003859 Key Organics / BIONET

methyl 7-chloro-1-benzothiophene-2-carboxylate

  • MDL number MFCD09027105
  • Cas Number : 550998-56-6
  • Molecular Weight : 226.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86 - 88
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97269

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GA-0933_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart