KEY128003863 Key Organics / BIONET

methyl 7-(trifluoromethyl)-1-benzothiophene-2-carboxylate

  • MDL number MFCD09027108
  • Cas Number : 550998-55-5
  • Molecular Weight : 260.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71 - 74
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97275

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GA-0942_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart