KEY128003872 Key Organics / BIONET

1-[(4-bromophenyl)methyl]-N,5-dimethyl-1H-1,2,3-triazole-4-carboxamide

  • MDL number MFCD09607957
  • Cas Number : 952182-54-6
  • Molecular Weight : 309.167

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186 dec
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97597

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/H-20173_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart