KEY128003878 Key Organics / BIONET

(3E)-6-chloro-3-(hydroxyimino)-3,4-dihydro-2H-1,4-benzoxazin-2-one

  • MDL number MFCD08689793
  • Cas Number : 937602-19-2
  • Molecular Weight : 212.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231 - 233
Purity 95
Storage temp. RT
Physical Form Brown to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97656

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/HA-0705_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart