KEY128003896 Key Organics / BIONET

N-amino-1-[N'-(3,4-dimethylphenyl)carbamimidamido]methanimidamide hydrochloride

  • MDL number MFCD09027118
  • Cas Number : 1171647-17-8
  • Molecular Weight : 256.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99566

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-160C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart