KEY128003913 Key Organics / BIONET

2-[4-(benzyloxy)phenyl]-1,3-thiazolidine

  • MDL number MFCD09027139
  • Cas Number : 937602-44-3
  • Molecular Weight : 271.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 134 - 136
Iupac Name 2-[4-(benzyloxy)phenyl]-1,3-thiazolidine
Inchi Code 1S/C16H17NOS/c1-2-4-13(5-3-1)12-18-15-8-6-14(7-9-15)16-17-10-11-19-16/h1-9,16-17H,10-12H2
Inchi Key FYAJTHNVORAUAP-UHFFFAOYSA-N

Description

Melting Point 134 - 136
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100815

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/KA-0727_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart