KEY128003964 Key Organics / BIONET

ethyl 4-(4-methoxybenzoyl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD09027191
  • Cas Number : 952182-69-3
  • Molecular Weight : 275.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 55 - 57
Iupac Name ethyl 4-(4-methoxybenzoyl)-3-isoxazolecarboxylate
Inchi Code 1S/C14H13NO5/c1-3-19-14(17)12-11(8-20-15-12)13(16)9-4-6-10(18-2)7-5-9/h4-8H,3H2,1-2H3
Inchi Key PNVDVDLKWPJHLF-UHFFFAOYSA-N

Description

Melting Point 55 - 57
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102070

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MA-0725_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart