KEY128004011 Key Organics / BIONET

5-nitro-2,3-dihydro-1H-indazol-3-one

  • MDL number MFCD00012418
  • Cas Number : 61346-19-8
  • Molecular Weight : 179.135

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 280 - 282
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96273

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EA-0717_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart