KEY128004038 Key Organics / BIONET

methyl 1,3-benzoxazole-5-carboxylate

  • MDL number MFCD08689687
  • Cas Number : 924869-17-0
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 110 - 111
Iupac Name methyl 1,3-benzoxazole-5-carboxylate
Inchi Code 1S/C9H7NO3/c1-12-9(11)6-2-3-8-7(4-6)10-5-13-8/h2-5H,1H3
Inchi Key VHLBJWCXFIGALN-UHFFFAOYSA-N

Description

Melting Point 110 - 111
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96318

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/EA-0853_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart