KEY128004046 Key Organics / BIONET

methyl 3-bromo-4-hydroxy-5-nitrobenzoate

  • MDL number MFCD08689690
  • Cas Number : 40258-72-8
  • Molecular Weight : 276.042

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131 - 133
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96328

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EA-0875_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart