KEY128004061 Key Organics / BIONET

N-methyl-3-(trifluoromethyl)benzene-1-sulfonamide

  • MDL number MFCD09817446
  • Cas Number : 882423-09-8
  • Molecular Weight : 239.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62 - 64
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96347

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EB-0727_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart