KEY128004080 Key Organics / BIONET

4-[(1E)-(hydroxyimino)methyl]-1,2-dihydropyrimidin-2-one

  • MDL number MFCD02685605
  • Cas Number : 7460-56-2
  • Molecular Weight : 139.114

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96676

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FA-0252_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart