KEY128004081 Key Organics / BIONET

ethyl (2E)-3-(4-tert-butylphenyl)-2-cyanoprop-2-enoate

  • MDL number MFCD04241676
  • Cas Number : 247099-46-3
  • Molecular Weight : 257.333

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67 - 69
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2394

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10N-904_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart