KEY128004083 Key Organics / BIONET

[3-(benzyloxy)-4-methoxyphenyl]methanol

  • MDL number MFCD00183283
  • Cas Number : 1860-60-2
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9584

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-0016_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart