KEY128004124 Key Organics / BIONET

methyl 6-formylpyridine-3-carboxylate

  • MDL number MFCD08275010
  • Cas Number : 10165-86-3
  • Molecular Weight : 165.148

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121 - 123
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94442

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0711_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart