KEY128004140 Key Organics / BIONET

3-(2-chloroethyl)-1-(4-methoxyphenyl)urea

  • MDL number MFCD02671069
  • Cas Number : 65536-40-5
  • Molecular Weight : 228.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156 - 158
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96738

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FA-0804_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart