KEY128004146 Key Organics / BIONET

tert-butyl 2H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate

  • MDL number MFCD08689720
  • Cas Number : 924869-27-2
  • Molecular Weight : 223.276

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127 - 128
Purity 95
Storage temp. Store under N2, Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96748

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FA-0815_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart